Orders LinkContactUsLinkComponent
Spider evaporators
Spider with cylindric flasks, SJ29/32, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 12×20 ml cylindrical flasks with 29,2/32 NS Joint
Katalogové číslo 531-1322
Dodavatel:   BUCHI
Spider evaporators
Katalogové číslo 531-1322
Dodavatel:  BUCHI
Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ29/32, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 12×20 ml cylindrical flasks with 29,2/32 NS Joint
Přejít na:
Price
81 300,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifikace
  • Popis:
    Spider with cylindric flasks, SJ29/32, glass, incl. 12 cylindric flasks, SJ14.5/23, 20 ml
  • Typ:
    For simultaneous distillation from 12×20 ml cylindrical flasks with 29,2/32 NS Joint