Orders LinkContactUsLinkComponent
Drying flasks
Drying flasks
1000 ml, 29/32 NS
Catalog # 531-1982
Supplier:   BUCHI
Drying flasks
Catalog # 531-1982
Supplier:  BUCHI
Drying flasks
Drying flasks
1000 ml, 29/32 NS
Price
4 260,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Made of Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For rotary evaporators Rotavapor R-210/215. Pear shape with NS 29/32 neck. Glassware A, V, C.
Specifications
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    1000 ml