Receiving flasks
50 ml, uncoated, KS 35/20
Catalog # 531-1831
Supplier: BUCHI
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for BÜCHI rotary evaporators. With spherical joint socket KS 35/20.
Specifications
- Zásuvka:KS 35/20
- Type:uncoated
- Kapacita:50 ml