Orders LinkContactUsLinkComponent
Extractor thimbles for solids
Extractor thimbles for solids
02, 100 ml, 45/40 NS
Catalog # 533-2151
Supplier:   Lenz Laborglas
Extractor thimbles for solids
Catalog # 533-2151
Supplier:  Lenz Laborglas
Extractor thimbles for solids
Extractor thimbles for solids
02, 100 ml, 45/40 NS
Price
3 250,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For glass inserts for solids.
Documents
Specifications
  • Extrakční objem:
    100 ml
  • Velikost objímky:
    45/40 NS
  • Sekvence:
    02