Cylindrical separating funnels
250 ml, 29/32 NS, 5 ml
Katalogové číslo 527-1103
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With graduation.
Documents
BSE TSE Glass Columns
Certifikáty BSE/TSE
Specifikace
- Velikost objímky:29/32 NS
- Kapacita:250 ml
- Dělení:5 ml