Erlenmeyer flasks with screw caps
250 ml, 145 mm, 85 mm, GL 32
Katalogové číslo 214-1210
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with DIN thread and red PBT closure.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and a white labelling area. Screw cap with PTFE coated seal.
Documents
BSE TSE Glass Columns
Certifikáty BSE/TSE
Specifikace
- Ø vněj.:85 mm
- Závit:GL 32
- Kapacita:250 ml
- Výška:145 mm