Stopcocks, one-way
08, 6,0** mm, 21,5 NS
Katalogové číslo 227-3138
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Documents
BSE TSE Glass Columns
Certifikáty BSE/TSE
Specifikace
- Ø otvoru:6,0** mm
- Sekvence:08
- Kohout:21,5 NS