Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
14,5 NS
  SCOT286433607
Stopcocks for separating funnels, DURAN®
  SCOT286433607
Stopcocks for separating funnels, DURAN®
14,5 NS
18 570,00 Kč
1 * 25 KS
DURAN®, borosilicate glass, with polished short funnel, without key.
  • Very good chemical resistance
  • High temperature resistance
For glass manipulators.
  • Kohout:
    14,5 NS