Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
Stopcocks for separating funnels, DURAN®
12,5 NS
Katalogové číslo SCOT286433401
Dodavatel:   DWK Life Sciences
Stopcocks for separating funnels, DURAN®
Katalogové číslo SCOT286433401
Dodavatel:  DWK Life Sciences
Stopcocks for separating funnels, DURAN®
12,5 NS
Přejít na:
Price
18 020,00 Kč
Přihlásit se pro dostupnost
1 * 25 KS
Product Details & Documents
DURAN®, borosilicate glass, with polished short funnel, without key.
  • Very good chemical resistance
  • High temperature resistance
For glass manipulators.
Specifikace
  • Kohout:
    12,5 NS