Orders LinkContactUsLinkComponent
ORP electrode solutions
01, ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 220 mV, 60 Ampoul
Catalog # SCOI285138373
Supplier:   SI Analytics
ORP electrode solutions
Catalog # SCOI285138373
Supplier:  SI Analytics
CAS Number:  
ORP electrode solutions
ORP electrode solutions
01, ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 220 mV, 60 Ampoul
CAS Number:  
Price
6 210,00 Kč
Log in for availability
1 * 60 Ampoul
Product Details & Documents
ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.
Specifications
Specifications
  • Bal.:
    60 Ampoul
  • Popis:
    ORP electrode solution, redox voltage (Pt/Ag/AgCl (KCl 3 mol/l)) of 220 mV
  • Sekvence:
    01
Specifications