Orders LinkContactUsLinkComponent
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous
07, Electrolyte solution, L 2114 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, 250 ml in DURAN® glass bottle, 250 mL
Katalogové číslo 662-6184
Dodavatel:   SI Analytics
Electrolyte solutions, aqueous
Katalogové číslo 662-6184
Dodavatel:  SI Analytics
Číslo CAS:  
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous
07, Electrolyte solution, L 2114 2 mol/l KNO3 + 0.001 mol/l KCl, for Ag combination electrodes, 250 ml in DURAN® glass bottle, 250 mL
Přejít na:
Číslo CAS:  
Price
1 100,00 Kč
Přihlásit se pro dostupnost
1 * 250 mL
Product Details & Documents
Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.
Specifikace
Specifikace
  • Bal.:
    250 mL
  • Popis:
    Electrolyte solution, L 2114 2 mol/l KNO<sub>3</sub> + 0.001 mol/l KCl, for Ag combination electrodes, 250 ml in DURAN® glass bottle
  • Sekvence:
    07