Orders LinkContactUsLinkComponent
Cables for Electrodes
Cables for Electrodes
11, Plug cable combination, B 10 X (electrode socket, 10 m single core cable upto 135 °C, free end), 10 m
Catalog # 662-2216
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2216
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
11, Plug cable combination, B 10 X (electrode socket, 10 m single core cable upto 135 °C, free end), 10 m
Price
2 130,00 Kč
Log in for availability
1 * 1 KS
Specifications
  • Popis:
    Plug cable combination, B 10 X (electrode socket, 10 m single core cable upto 135 °C, free end)
  • Délka:
    10 m
  • Sekvence:
    11