Electrode Cables
19, S-connector, socket for cable extension, 1 m
Katalogové číslo SCOI285121616
Dodavatel: SI Analytics
Přejít na:
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Documents
Specifikace
- Popis:S-connector, socket for cable extension
- Délka:1 m
- Sekvence:19