Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
13, DIN connector for Schott, WTW pH meters, 2 m
Catalog # SCOI285120225
Supplier:   SI Analytics
Electrode Cables
Catalog # SCOI285120225
Supplier:  SI Analytics
Electrode Cables
Electrode Cables
13, DIN connector for Schott, WTW pH meters, 2 m
Přejít na:
Price
2 280,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Specifikace
  • Popis:
    DIN connector for Schott, WTW pH meters
  • Délka:
    2 m
  • Sekvence:
    13