Reference electrodes
Reference electrodes with free end, flanged cap, low maintatence, Ag/AgCl reference system, 9801/85, −5...+50 °C, Sklo, Paste, Ceramic, 12×120 mm, 5 m fixed cable
SCOI285117856
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
- Připojení:5 m fixed cable
- Ø×D:12×120 mm
- Type:9801/85
- Popis:Reference electrodes with free end, flanged cap, low maintatence, Ag/AgCl reference system
- Materiál:Sklo
- Diafragma:Ceramic
- Elektrolyt:Paste
- Tepl.:−5...+50 °C