Orders LinkContactUsLinkComponent
Lightlines
Lightlines
983, Lightline with adjustable front lens slit, 200×0,3 mm
Catalog # 631-0049
Lightlines
Catalog # 631-0049
Lightlines
Lightlines
983, Lightline with adjustable front lens slit, 200×0,3 mm
Price
37 380,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Popis:
    Lightline with adjustable front lens slit, 200×0,3 mm
  • Sekvence:
    983