Orders LinkContactUsLinkComponent
Microscopy slides
Microscopy slides
Slides with cut edges, Bílá/průhledná, 76×26 mm
Catalog # BRND474701
Supplier:   Brand
Microscopy slides
Catalog # BRND474701
Supplier:  Brand
Microscopy slides
Microscopy slides
Slides with cut edges, Bílá/průhledná, 76×26 mm
Price
4 830,00 Kč
Log in for availability
1 * 2.500 KS
Product Details & Documents
Microscopy slides with ground edges to reduce the risk of injury.
Specifications
  • Barva:
    Bílá/průhledná
  • Popis:
    Slides with cut edges
  • D׊:
    76×26 mm