Orders LinkContactUsLinkComponent
Paper scissors
Paper scissors
Sharp/sharp, 225 mm
Catalog # BOCH4162
Supplier:   BOCHEM
Paper scissors
Catalog # BOCH4162
Supplier:  BOCHEM
Paper scissors
Paper scissors
Sharp/sharp, 225 mm
Price
465,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Straight paper scissors made of stainless steel.
Specifications
  • Type:
    Sharp/sharp
  • Délka:
    225 mm