Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
01, Sharp/blunt, 100 mm
Catalog # BOCH4000
Supplier:   BOCHEM
Laboratory scissors, straight
Catalog # BOCH4000
Supplier:  BOCHEM
Laboratory scissors, straight
Laboratory scissors, straight
01, Sharp/blunt, 100 mm
Price
216,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Straight scissors made of stainless steel.
 : These scissors are not recommended for autoclaving.
Specifications
  • Type:
    Sharp/blunt
  • Délka:
    100 mm
  • Sekvence:
    01