Orders LinkContactUsLinkComponent
Transport containers
Transport containers
05, Slide mailers, PP, 82×17×29 mm, 5
Katalogové číslo 631-9780
Dodavatel:   DELTALAB
Transport containers
Katalogové číslo 631-9780
Dodavatel:  DELTALAB
Transport containers
Transport containers
05, Slide mailers, PP, 82×17×29 mm, 5
Přejít na:
Price
223,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.
Documents
Specifikace
  • Počet sklíček:
    5
  • Popis:
    Slide mailers
  • Materiál:
    PP
  • Sekvence:
    05
  • Š×H×V:
    82×17×29 mm