Orders LinkContactUsLinkComponent
Pipette controller, Pi-Pump
Pipette controller Pi-Pump, red, for 25 ml pipettes
Katalogové číslo 612-1927
Dodavatel:   KARTELL
Pipette controller, Pi-Pump
Katalogové číslo 612-1927
Dodavatel:  KARTELL
Pipette controller, Pi-Pump
Pipette controller, Pi-Pump
Pipette controller Pi-Pump, red, for 25 ml pipettes
Přejít na:
Price
473,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Pipetting aid for all plastic and glass pipettes.
  • Suction and dispensing achieved by rotating the wheel
  • Liquid evacuated by pressing the lever
Specifikace
  • Popis:
    Pipette controller Pi-Pump, red, for 25 ml pipettes