Orders LinkContactUsLinkComponent
Column end plugs
Column end plugs
01, Column end plug, white
Katalogové číslo 554-0759
Dodavatel:   VICI JOUR RESEARCH
Column end plugs
Katalogové číslo 554-0759
Dodavatel:  VICI JOUR RESEARCH
Column end plugs
Column end plugs
01, Column end plug, white
Přejít na:
Price
477,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
These nylon column end plugs fit all columns with 10-32 threads. The long design and threading down to the ferrule ensure safe closing of valuable columns.
  • Available with a 5/16" wrench flat
  • Easy removal
Specifikace
  • Popis:
    Column end plug, white
  • Sekvence:
    01