Orders LinkContactUsLinkComponent
Column end plugs
03, Column end plug, red
Catalog # 555-3595
Supplier:   VICI JOUR RESEARCH
Column end plugs
Catalog # 555-3595
Supplier:  VICI JOUR RESEARCH
Column end plugs
Column end plugs
03, Column end plug, red
Price
477,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
These nylon column end plugs fit all columns with 10-32 threads. The long design and threading down to the ferrule ensure safe closing of valuable columns.
  • Available with a 5/16" wrench flat
  • Easy removal
Specifications
  • Popis:
    Column end plug, red
  • Sekvence:
    03