Dry ice machines, SnowPack
Snowpack® 50 - gas outflow time/tablet (min):~ 0,5, 50×30 mm
Katalogové číslo 474-3110
Dodavatel: BURKLE
Přejít na:
Product Details & Documents
SnowPack enables the user to obtain a −79 °C dry ice tablet within minutes, without complicated technology and without electricity. Can be installed on any commercial CO₂-bottle.
- Ideal coolant for transporting
- Dry ice is odourless and easy to handle
Documents
SnowPack
Videa
Specifikace
- Popis:Snowpack® 50 - gas outflow time/tablet (min):~ 0,5
- Ø×H:50×30 mm