Tripod stands
05, 140×220 mm
Katalogové číslo 451-9103
Dodavatel: USBECK Laborgeräte
Přejít na:
Product Details & Documents
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
Specifikace
- Vnitř. Ø×V:140×220 mm
- Sekvence:05