Orders LinkContactUsLinkComponent
Pestles
Pestles
13, 28 mm, 125 mm, 410-9121, 410-9111
Catalog # 410-9128
Pestles
Catalog # 410-9128
Pestles
Pestles
13, 28 mm, 125 mm, 410-9121, 410-9111
Price
426,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Porcelain with glazed handle, glazed or unglazed grinding surface.
Certifications: Manufactured to DIN 12906.
Specifications
  • Určeno pro:
    410-9121, 410-9111
  • Ø hlavy:
    28 mm
  • Délka:
    125 mm
  • Sekvence:
    13