Microplates, 96-well
04, U-bottom, clear, sterile, 40 - 280 µl, 96, +, 323 µl
Katalogové číslo 391-3311
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Documents
Specifikace
- Poč. jamek:96
- Doporučený pracovní objem:40 - 280 µl
- Sterilní:+
- Popis:U-bottom, clear, sterile
- Sekvence:04
- Celkový objem:323 µl