QualiSampler sets
QualiSampler set with LiquiSampler, ViscoSampler, sample bottles (5), a cleaning brush and a carrying case, PTFE
Catalog # 331-2181
Supplier: BURKLE
Jump to:
Product Details & Documents
Complete handy set combining the LiquiSampler and the ViscoSampler with practical accessories.
Delivery information: Set consists of LiquiSampler 100 cm, ViscoSampler 100 cm, case, 5 sample bottles, cleaning brush.
Specifications
- Popis:QualiSampler set with LiquiSampler, ViscoSampler, sample bottles (5), a cleaning brush and a carrying case
- Materiál:PTFE