Orders LinkContactUsLinkComponent
Forceps
Forceps
01, Forceps, College, Angled, smooth, fine tips, 150 mm
Catalog # 232-1260
Supplier:   HAMMACHER KARL
Forceps
Catalog # 232-1260
Supplier:  HAMMACHER KARL
Forceps
Forceps
01, Forceps, College, Angled, smooth, fine tips, 150 mm
Price
1 560,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Type:
    Angled, smooth, fine tips
  • Popis:
    Forceps, College
  • Délka:
    150 mm
  • Sekvence:
    01