Double spatulas, micro
130 mm, 5 mm
231-2261
: RSG
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
- Both ends have an angled spoon
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
- Délka:130 mm
- Šířka čepele:5 mm