Double spatulas, micro
150 mm, 6 mm
Katalogové číslo 231-2239
Dodavatel: RSG
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Very flexible blades
Documents
RSGA032.150_TDS_EN
Datasheet
Specifikace
- Délka:150 mm
- Šířka čepele:6 mm