Orders LinkContactUsLinkComponent
Double spatulas, micro
150 mm, 5 mm
Catalog # 231-2233
Supplier:   RSG
Double spatulas, micro
Catalog # 231-2233
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro
150 mm, 5 mm
Price
1 050,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades
Specifications
  • Délka:
    150 mm
  • Šířka čepele:
    5 mm