Orders LinkContactUsLinkComponent
Connectors
03, Fittings, external thread for pipes and valves, 1"
Katalogové číslo 229-0061
Dodavatel:   SEMADENI
Connectors
Katalogové číslo 229-0061
Dodavatel:  SEMADENI
Connectors
Connectors
03, Fittings, external thread for pipes and valves, 1"
Přejít na:
Price
868,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Connectors for easy installation on any plastic containers by means of counter screw.
  • Male thread for fittings or hose connection
  • Good chemical resistance to acids and solvents
Specifikace
  • Závit:
    1"
  • Popis:
    Fittings, external thread for pipes and valves
  • Sekvence:
    03