Jars with overlapping lid
08, 46 mm, 16 mm
Catalog # 216-1265
Supplier: SEMADENI
Jump to:
Product Details & Documents
Jars, made of PS, clear, with closing lid.
- Good chemical resistance
- Easy to clean
Suitable for ointments, powders, and granules.
Specifications
- Ø vněj.:46 mm
- Výška:16 mm
- Sekvence:08