Orders LinkContactUsLinkComponent
Draining racks, with drip pan
Draining racks, with drip pan
02, Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm), 420×160×610 mm
Katalogové číslo BOCH10130
Dodavatel:   BOCHEM
Draining racks, with drip pan
Katalogové číslo BOCH10130
Dodavatel:  BOCHEM
Draining racks, with drip pan
Draining racks, with drip pan
02, Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm), 420×160×610 mm
Přejít na:
Price
3 990,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These PA coated wire draining racks are suitable for test tubes and flasks.
  • With drip pan
  • Good chemical resistance
  • Good temperature resistance
Specifikace
  • Popis:
    Draining rack, wth 60 rods (95 mm) and 5 arches (2×110 mm and 3×70 mm)
  • Sekvence:
    02
  • Š×H×V:
    420×160×610 mm