Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
2000 ml, 29/32 NS, 50 ml
Katalogové číslo 527-1995
Dodavatel:   Lenz Laborglas
Separating funnels, Squibb
Katalogové číslo 527-1995
Dodavatel:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
2000 ml, 29/32 NS, 50 ml
Přejít na:
Price
3 620,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.
Documents
Specifikace
  • Velikost objímky:
    29/32 NS
  • Kapacita:
    2000 ml
  • Dělení:
    50 ml