Orders LinkContactUsLinkComponent
Spray bottles
Spray bottles
01, 400 ml, Spray bottle, Bílá
Catalog # VITL53510
Supplier:   VITLAB
Spray bottles
Catalog # VITL53510
Supplier:  VITLAB
Spray bottles
Spray bottles
01, 400 ml, Spray bottle, Bílá
Price
1 490,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
White or translucent made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle, which can be regulated from the finest mist (nebulising) to a precise liquid jet. Range: approx. 3 to 4 metres. Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography.
Specifications
  • Kapacita:
    400 ml
  • Barva:
    Bílá
  • Popis:
    Spray bottle
  • Sekvence:
    01