Measuring scoops
2 ml, 60 mm
Catalog # 231-0326
Supplier: VITLAB
Jump to:
Product Details & Documents
PP, white. With flat, chamfered ends and reinforced handle.
- Excellent chemical resistance, robust
- Heat resistant up to +130 °C, autoclavable
- The capacity of the scoop is stamped into the handle for easy reading
Documents
BSE TSE VITL39794.pdf BSE TSE VITL39894.pdf
BSE/TSE Certificates
Specifications
- Kapacita:2 ml
- Délka:60 mm