Orders LinkContactUsLinkComponent
Dishes
Dishes
03, 1 l, 90 mm, 175 mm
Catalog # 216-1241
Supplier:   RSG
Dishes
Catalog # 216-1241
Supplier:  RSG
Dishes
Dishes
03, 1 l, 90 mm, 175 mm
Price
655,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
  • With folded edge; fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
Round multipurpose dishes for a wide range of uses.
 : Each dish set (Cat. No. 216-1249) comprises one of each 1, 2 and 3 litre dishes.
Specifications
  • Ø vněj.:
    175 mm
  • Kapacita:
    1 l
  • Výška:
    90 mm
  • Sekvence:
    03