Orders LinkContactUsLinkComponent
Forceps
Forceps
Curved, pointed, 145 mm
Catalog # 232-2133
Supplier:   RSG
Forceps
Catalog # 232-2133
Supplier:  RSG
Forceps
Forceps
Curved, pointed, 145 mm
Price
278,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
Documents
Specifications
  • Type:
    Curved, pointed
  • Délka:
    145 mm