Support rings
Heavy support ring, with clamp, open, with rubber coating, 130 mm
RSGA284.130
: RSG
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
- Ø vnitř.:130 mm
- Popis:Heavy support ring, with clamp, open, with rubber coating