Support rings
Heavy support ring, with clamp, open, with rubber coating, 50 mm
RSGA284.050
: RSG
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
- Ø vnitř.:50 mm
- Popis:Heavy support ring, with clamp, open, with rubber coating