Support rings
Heavy support ring, with rod, open, with rubber coating, 160 mm
Katalogové číslo RSGA283.160
Dodavatel: RSG
Přejít na:
Product Details & Documents
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
Specifikace
- Ø vnitř.:160 mm
- Popis:Heavy support ring, with rod, open, with rubber coating