Support rings
Heavy support ring, with clamp, 70 mm
Catalog # 241-0321
Supplier: RSG
Jump to:
Product Details & Documents
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
Specifications
- Ø vnitř.:70 mm
- Popis:Heavy support ring, with clamp