Orders LinkContactUsLinkComponent
Spoon spatulas
250 mm
Catalog # 231-0439
Supplier:   RSG
Spoon spatulas
Catalog # 231-0439
Supplier:  RSG
Spoon spatulas
Spoon spatulas
250 mm
Price
2 330,00 Kč
Log in for availability
1 * 5 KS
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon spatula for a wide range of uses.
  • With round handle
  • Very good thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Délka:
    250 mm