Spoon spatulas
180 mm
Catalog # 231-0437
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon spatula for a wide range of uses.
- With round handle
- Very good thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Documents
RSGA210.150_TDS_EN RSGA210.180_TDS_EN
Datasheet
Specifications
- Délka:180 mm