Spoon spatulas
150 mm
Katalogové číslo 231-0436
Dodavatel: RSG
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoon spatula for a wide range of uses.
- With round handle
- Very good thermal resistance, autoclavable
- Good chemical resistance
- Quick and easy to clean
Documents
RSGA210.150_TDS_EN RSGA210.180_TDS_EN
Technický list
Specifikace
- Délka:150 mm