Orders LinkContactUsLinkComponent
General purpose scissors
General purpose scissors
01, Straight, 150 mm
Catalog # RSGA109.160
Supplier:   RSG
General purpose scissors
Catalog # RSGA109.160
Supplier:  RSG
General purpose scissors
General purpose scissors
01, Straight, 150 mm
Price
1 310,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Ice-tempered scissors with stainless steel blades and plastic covered handle.
Specifications
  • Type:
    Straight
  • Délka:
    150 mm
  • Sekvence:
    01