Orders LinkContactUsLinkComponent
Small flanges with hose nozzle
Small flanges with hose nozzle
13, Small flanges, DN 40, Hliník, 15 mm
Katalogové číslo 185-2259
Dodavatel:   VACUUBRAND
Small flanges with hose nozzle
Katalogové číslo 185-2259
Dodavatel:  VACUUBRAND
Small flanges with hose nozzle
Small flanges with hose nozzle
13, Small flanges, DN 40, Hliník, 15 mm
Přejít na:
Price
1 630,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These flanges are made from aluminium or plastic.
  • Fast, easy assembly and disassembly to/from pumps
  • Safe, vacuum tight connection with optional centring and clamping rings
  • A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.
Specifikace
  • Hadice int.Ø:
    15 mm
  • Popis:
    Small flanges, DN 40
  • Materiál:
    Hliník
  • Sekvence:
    13