Small flanges with hose nozzle
11, Small flanges, DN 40, Hliník, 8 mm
185-2257
These flanges are made from aluminium or plastic.
- Fast, easy assembly and disassembly to/from pumps
- Safe, vacuum tight connection with optional centring and clamping rings
- A wide range of flanges with hose nozzles for various internal hose diameters are available
For connecting pumps to vacuum hoses.
- Hadice int.Ø:8 mm
- Popis:Small flanges, DN 40
- Materiál:Hliník
- Sekvence:11